missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic Acid, TCI America™

$531.67 - $1,591.98

Chemical Identifiers

CAS 178306-52-0
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.3
MDL Number MFCD11113284
InChI Key RQJWOLFMWKZKCJ-CQSZACIVSA-N
PubChem CID 15380898
IUPAC Name (2S)-2-hydroxy-3-methoxy-3,3-diphenylpropanoic acid
SMILES COC(C1=CC=CC=C1)(C2=CC=CC=C2)C(C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H15685G View Documents TCI America
H15685G
5g Each for $1,591.98
 
H15681G View Documents TCI America
H15681G
1g Each for $531.67
 
Specifications

Chemical Identifiers

178306-52-0
272.3
RQJWOLFMWKZKCJ-CQSZACIVSA-N
(2S)-2-hydroxy-3-methoxy-3,3-diphenylpropanoic acid
C16H16O4
MFCD11113284
15380898
COC(C1=CC=CC=C1)(C2=CC=CC=C2)C(C(=O)O)O

Specifications

White
5g
C16H16O4
RQJWOLFMWKZKCJ-CQSZACIVSA-N
(2S)-2-hydroxy-3-methoxy-3,3-diphenylpropanoic acid
15380898
Crystal-Powder at 20°C
(S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic Acid
120°C
178306-52-0
MFCD11113284
COC(C1=CC=CC=C1)(C2=CC=CC=C2)C(C(=O)O)O
272.3
272.30
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : No