Please login to your online account to display your discounted pricing

(S)-2-Hydroxy-3-buten-1-yl p-tosylate, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC411970050

 View more versions of this product

Catalog No. AC411970050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder or Crystals
Assay ≥98.5% (HPLC)
Formula Weight 242.3
Quantity 5g
Color White to beige
Specific Rotation Condition − 78.50 (20.00°C c=1.13 MeOH)
Melting Point 57° to 63°C
Specific Rotation −7° to −8.5° (20°C, 589nm) (c=1.13, MeOH)
Assay Percent Range 99%
Molecular Formula C11H14O4S
Chemical Name or Material (S)-2-Hydroxy-3-buten-1-yl p-tosylate
Packaging Glass bottle
CAS 133095-74-6
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (S)-2-Hydroxy-3-buten-1-yl p-tosylate
Assay ≥98.5% (HPLC)
Assay Percent Range 99%
Molecular Formula C11H14O4S
Formula Weight 242.3