Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-2-Hexyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material (S)-(+)-2-Hexyl isocyanate
CAS 745783-78-2
Molecular Formula C7H13NO
Formula Weight 127.19
MDL Number MFCD05664080
UN Number UN3080
Beilstein 8253640
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2023503

  SDS
alfa aesar™
L2023503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 127.19
Chemical Name or Material (S)-(+)-2-Hexyl isocyanate
Assay 97%
Molecular Formula C7H13NO
MDL Number MFCD05664080
CAS 745783-78-2
UN Number UN3080
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H330-H334-H226-H315-H319-H335
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 8253640
Quantity 1g
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H330-H334-H226-H315-H319-H335
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C