missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-2-Hexanol, 98%

Manufacturer:  Alfa Aesar™ L10401MD

 View more versions of this product

Catalog No. AAL10401MD



Description

Description

Applications
S)-(+)-2-Hexanol is the S enantiomer used in the preparation of key intermediates for model studies in the total synthesis of antivirally active glycolipid cycloviracin B1.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

(S)-(+)-2-Hexanol
+10.5° (Neat)
250mg
C6H14O
UN2282
s-+-2-hexanol, s-2-hexanol, s-+-2-hexanol, 2s-hexan-2-ol, 2-hexanol #, s-2-hexanol, pubchem6732, 2s-2-hexanol, s-hexan-2-ol, s +-2-hexanol
CCCCC(C)O
102.177
102.18
45°C (113°F)
137°C to 138°C
52019-78-0
MFCD00065955
1718997
QNVRIHYSUZMSGM-LURJTMIESA-N
(2S)-hexan-2-ol
638097
98%

Chemical Identifiers

52019-78-0
102.177
QNVRIHYSUZMSGM-LURJTMIESA-N
638097
CCCCC(C)O
C6H14O
MFCD00065955
s-+-2-hexanol, s-2-hexanol, s-+-2-hexanol, 2s-hexan-2-ol, 2-hexanol #, s-2-hexanol, pubchem6732, 2s-2-hexanol, s-hexan-2-ol, s +-2-hexanol
(2S)-hexan-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.

GHS P Statement
P261-P280f
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective clothing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXANOLS

Documents