Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-2-Heptanol, 98%

Chemical Identifiers

Chemical Name or Material (S)-(+)-2-Heptanol
CAS 6033-23-4
Formula Weight 116.2
MDL Number MFCD00065003
UN Number UN1987
Merck Index 14,4662
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3379506

  SDS
alfa aesar™
3379506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3379503

  SDS
alfa aesar™
3379503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA33795MD

  SDS
alfa aesar™
33795MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.2
Chemical Name or Material (S)-(+)-2-Heptanol
Density 0.815g/mL
Physical Form Liquid
Assay 98%
MDL Number MFCD00065003
CAS 6033-23-4
UN Number UN1987
Merck Index 14,4662
Health Hazard 1 Danger
Refractive Index 1.4210
Health Hazard 2 GHS H Statement
H311-H226-H319
Toxic in contact with skin.
Flammable liquid and vapor.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 160° to 162°C
Quantity 5g
Flash Point 59° (138°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H311-H226-H319
Toxic in contact with skin.
Flammable liquid and vapor.
Causes serious eye irritation.