missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-2-(Fmoc-amino)-4-aminobutyric acid, 98%

$47.04 - $462.64

Chemical Identifiers

CAS 161420-87-7
Molecular Formula C19H20N2O4
Molecular Weight (g/mol) 340.379
MDL Number MFCD00237017
InChI Key ZZDRDGKSMGGBDI-KRWDZBQOSA-N
Synonym fmoc-dab-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoic acid, fmoc-l-2,4-diaminobutyric acid, 2s-4-amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, s-n2-fmoc-2,4-diaminobutyric acid, ambotzfaa1456, na-fmoc-l-2,4-diaminobutyric acid, n-a-fmoc-l-2,4-diaminobutyric acid, 2s-2-9h-fluorene-9-ylmethoxycarbonylamino-4-aminobutanoic acid, 2s-4-amino-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid
PubChem CID 7019708
IUPAC Name (2S)-4-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCN)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66097MD View Documents Alfa Aesar™
H66097MD
250mg Each for $47.04
 
AAH6609703 View Documents Alfa Aesar™
H6609703
1g Each for $124.14
 
AAH6609706 View Documents Alfa Aesar™
H6609706
5g Each for $462.64
 
Specifications

Chemical Identifiers

161420-87-7
340.379
ZZDRDGKSMGGBDI-KRWDZBQOSA-N
7019708
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCN)C(=O)O
C19H20N2O4
MFCD00237017
fmoc-dab-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoic acid, fmoc-l-2,4-diaminobutyric acid, 2s-4-amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, s-n2-fmoc-2,4-diaminobutyric acid, ambotzfaa1456, na-fmoc-l-2,4-diaminobutyric acid, n-a-fmoc-l-2,4-diaminobutyric acid, 2s-2-9h-fluorene-9-ylmethoxycarbonylamino-4-aminobutanoic acid, 2s-4-amino-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid
(2S)-4-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid

Specifications

White
161420-87-7
MFCD00237017
ZZDRDGKSMGGBDI-KRWDZBQOSA-N
(2S)-4-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
7019708
Powder
(S)-2-(Fmoc-amino)-4-aminobutyric acid
250mg
C19H20N2O4
fmoc-dab-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoic acid, fmoc-l-2,4-diaminobutyric acid, 2s-4-amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, s-n2-fmoc-2,4-diaminobutyric acid, ambotzfaa1456, na-fmoc-l-2,4-diaminobutyric acid, n-a-fmoc-l-2,4-diaminobutyric acid, 2s-2-9h-fluorene-9-ylmethoxycarbonylamino-4-aminobutanoic acid, 2s-4-amino-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCN)C(=O)O
340.379
340.38
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C