missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-2-Diphenylphosphino-1-phenylethylamine, 97+%

$133.54 - $408.66

Chemical Identifiers

CAS 1103533-85-2
Molecular Formula C20H20NP
Molecular Weight (g/mol) 305.361
MDL Number MFCD17013989
InChI Key OTNYDYNCOCRMDG-HXUWFJFHSA-N
Synonym s-2-diphenylphosphino-1-phenylethanamine, s-2-diphenylphosphino-1-phenylethylamine, s-2-diphenylphosphino-1-phenylethylamine, 2s-2-amino-2-phenylethyl diphenylphosphane, alphachiron 1112421a940, s-beta-aminophenethyl diphenylphosphine, s-2-diphenylphosphanyl-1-phenylethan-1-amine, 1s-2-diphenylphosphanyl-1-phenylethan-1-amine, s-2-diphenylphosphino-1-phenylethylamine,min
PubChem CID 70861903
IUPAC Name (1S)-2-diphenylphosphanyl-1-phenylethanamine
SMILES C1=CC=C(C=C1)C(CP(C2=CC=CC=C2)C3=CC=CC=C3)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60404MD View Documents Alfa Aesar™
H60404MD
250mg Each for $133.54
 
AAH6040403 View Documents Alfa Aesar™
H6040403
1g Each for $408.66
 
Specifications

Chemical Identifiers

1103533-85-2
305.361
OTNYDYNCOCRMDG-HXUWFJFHSA-N
70861903
C1=CC=C(C=C1)C(CP(C2=CC=CC=C2)C3=CC=CC=C3)N
C20H20NP
MFCD17013989
s-2-diphenylphosphino-1-phenylethanamine, s-2-diphenylphosphino-1-phenylethylamine, s-2-diphenylphosphino-1-phenylethylamine, 2s-2-amino-2-phenylethyl diphenylphosphane, alphachiron 1112421a940, s-beta-aminophenethyl diphenylphosphine, s-2-diphenylphosphanyl-1-phenylethan-1-amine, 1s-2-diphenylphosphanyl-1-phenylethan-1-amine, s-2-diphenylphosphino-1-phenylethylamine,min
(1S)-2-diphenylphosphanyl-1-phenylethanamine

Specifications

250mg
C20H20NP
s-2-diphenylphosphino-1-phenylethanamine, s-2-diphenylphosphino-1-phenylethylamine, s-2-diphenylphosphino-1-phenylethylamine, 2s-2-amino-2-phenylethyl diphenylphosphane, alphachiron 1112421a940, s-beta-aminophenethyl diphenylphosphine, s-2-diphenylphosphanyl-1-phenylethan-1-amine, 1s-2-diphenylphosphanyl-1-phenylethan-1-amine, s-2-diphenylphosphino-1-phenylethylamine,min
C1=CC=C(C=C1)C(CP(C2=CC=CC=C2)C3=CC=CC=C3)N
305.361
305.35
(S)-2-Diphenylphosphino-1-phenylethylamine
1103533-85-2
MFCD17013989
OTNYDYNCOCRMDG-HXUWFJFHSA-N
(1S)-2-diphenylphosphanyl-1-phenylethanamine
70861903
≥97%