missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-2-Diphenylphosphino-1-phenylethylamine, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H60404MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(S)-2-Diphenylphosphino-1-phenylethylamine | |
C20H20NP | |
250 mg | |
s-2-diphenylphosphino-1-phenylethanamine, s-2-diphenylphosphino-1-phenylethylamine, 2s-2-amino-2-phenylethyl diphenylphosphane, alphachiron 1112421a940, s-beta-aminophenethyl diphenylphosphine, s-2-diphenylphosphanyl-1-phenylethan-1-amine, 1s-2-diphenylphosphanyl-1-phenylethan-1-amine, s-2-diphenylphosphino-1-phenylethylamine,min | |
N[C@H](CP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 | |
305.36 | |
305.35 |
1103533-85-2 | |
MFCD17013989 | |
Air Sensitive | |
OTNYDYNCOCRMDG-GNLPSFAGNA-N | |
(1S)-2-diphenylphosphanyl-1-phenylethanamine | |
70861903 | |
≥97% |
Chemical Identifiers
1103533-85-2 | |
305.36 | |
OTNYDYNCOCRMDG-GNLPSFAGNA-N | |
70861903 | |
N[C@H](CP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
C20H20NP | |
MFCD17013989 | |
s-2-diphenylphosphino-1-phenylethanamine, s-2-diphenylphosphino-1-phenylethylamine, 2s-2-amino-2-phenylethyl diphenylphosphane, alphachiron 1112421a940, s-beta-aminophenethyl diphenylphosphine, s-2-diphenylphosphanyl-1-phenylethan-1-amine, 1s-2-diphenylphosphanyl-1-phenylethan-1-amine, s-2-diphenylphosphino-1-phenylethylamine,min | |
(1S)-2-diphenylphosphanyl-1-phenylethanamine |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only