missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-2-Dibenzylamino-3-phenyl-1-propanol, 99%

Manufacturer:  Alfa Aesar™ L1391603

 View more versions of this product

Catalog No. AAL1391603



Specifications

Specifications

(S)-(+)-2-Dibenzylamino-3-phenyl-1-propanol
72°C to 74°C
111060-52-7
MFCD00191984
s-+-2-dibenzylamino-3-phenyl-1-propanol, s-2-dibenzylamino-3-phenylpropan-1-ol, s-+-2-dibenzylamino-3-phenyl-1-propanol, 2s-2-dibenzylamino-3-phenylpropan-1-ol, dibenzyl phenylalaninol, n,n-dibenzyl-l-phenylalaninol, 2-s-dibenzylamino-3-phenylpropan-1-ol, s-beta-dibenzylamino benzene-1-propanol, s--2-dibenzylamino-3-phenyl-1-propanol, benzenepropanol,b-bis phenylmethyl amino-, bs
C1=CC=C(C=C1)CC(CO)N(CC2=CC=CC=C2)CC3=CC=CC=C3
331.459
331.46
+8.5° (c=4 in Methanol)
1g
C23H25NO
7253058
ZXNVOFMPUPOZDF-QHCPKHFHSA-N
(2S)-2-(dibenzylamino)-3-phenylpropan-1-ol
7019180
99%

Chemical Identifiers

111060-52-7
331.459
ZXNVOFMPUPOZDF-QHCPKHFHSA-N
7019180
C1=CC=C(C=C1)CC(CO)N(CC2=CC=CC=C2)CC3=CC=CC=C3
C23H25NO
MFCD00191984
s-+-2-dibenzylamino-3-phenyl-1-propanol, s-2-dibenzylamino-3-phenylpropan-1-ol, s-+-2-dibenzylamino-3-phenyl-1-propanol, 2s-2-dibenzylamino-3-phenylpropan-1-ol, dibenzyl phenylalaninol, n,n-dibenzyl-l-phenylalaninol, 2-s-dibenzylamino-3-phenylpropan-1-ol, s-beta-dibenzylamino benzene-1-propanol, s--2-dibenzylamino-3-phenyl-1-propanol, benzenepropanol,b-bis phenylmethyl amino-, bs
(2S)-2-(dibenzylamino)-3-phenylpropan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents