missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-2-Cyclohexylsuccinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 362092500

 View more versions of this product

Catalog No. AC362092500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

(S)-2-Cyclohexylsuccinic acid-1-methyl ester
250mg
213270-44-1
100.0
Authentic
Crystalline Chunks
95%
214.26
(S)-3-Cyclohexyl-3-(methoxycarbonyl)propanoic acid
White to beige
95%, (98% e.e.)
94.0
97.5% min. (GC)
Glass bottle
≥94%
C11H18O4
MFCD01091027

Chemical Identifiers

213270-44-1
100.0
C11H18O4
MFCD01091027
(S)-2-Cyclohexylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
94.0
95%
214.26
(S)-3-Cyclohexyl-3-(methoxycarbonyl)propanoic acid
≥94%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA