Please login to your online account to display your discounted pricing

(S)-2-Cyclohexylsuccinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  362090010

 View more versions of this product

Catalog No. AC362090010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 214.26
Physical Form Crystalline Chunks
Quantity 1g
Color White to beige
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Synonym (S)-3-Cyclohexyl-3-(methoxycarbonyl)propanoic acid
MDL Number MFCD01091027
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (GC)
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Molecular Formula C11H18O4
CAS 213270-44-1
Chemical Name or Material (S)-2-Cyclohexylsuccinic acid-1-methyl ester
Packaging Glass bottle

Specifications

Chemical Name or Material (S)-2-Cyclohexylsuccinic acid-1-methyl ester
Name Note 95%, (98% e.e.)
CAS 213270-44-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H18O4
Formula Weight 214.26
MDL Number MFCD01091027
Synonym (S)-3-Cyclohexyl-3-(methoxycarbonyl)propanoic acid