missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-2-Cyclohexylsuccinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™

$1,251.08 - $1,251.08

Chemical Identifiers

CAS 213270-44-1
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C11H18O4
Formula Weight 214.26
MDL Number MFCD01091027
Synonym (S)-3-Cyclohexyl-3-(methoxycarbonyl)propanoic acid
Chemical Name or Material (S)-2-Cyclohexylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC362092500 SDS Thermo Fisher Scientific
362092500
Glass bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC362090010 SDS Thermo Fisher Scientific
362090010
Glass bottle 1g Each for $1,251.08
Specifications Description & Specifications

Chemical Identifiers

213270-44-1
100.0
C11H18O4
MFCD01091027
(S)-2-Cyclohexylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
94.0
95%
214.26
(S)-3-Cyclohexyl-3-(methoxycarbonyl)propanoic acid
≥94%

Specifications

214.26
250mg
94.0
Glass bottle
C11H18O4
95%
97.5% min. (GC)
(S)-2-Cyclohexylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
Crystalline Chunks
White to beige
100.0
(S)-3-Cyclohexyl-3-(methoxycarbonyl)propanoic acid
213270-44-1
MFCD01091027
Authentic
≥94%

Safety and Handling

TSCA : TSCA