Please login to your online account to display your discounted pricing

(S)-2-(Cyclohexylmethyl)succinic acid-1-methyl ester, 95%, 98% e.e., Acros Organics


Quantity 250mg
Packaging Glass Ampule with screw-cap

Manufacturer: thermo fisher scientific  362082500

 View more versions of this product

Catalog No. AC362082500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Viscous Liquid
Formula Weight 228.28
Quantity 250mg
Color Clear colorless to pale yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass Ampule with screw-cap
Assay Percent Range 95%
MDL Number MFCD01311762
Chemical Name or Material (S)-2-(Cyclohexylmethyl)succinic acid-1-methyl ester
Infrared Spectrum Authentic
Name Note 95%, (98% e.e.)
Molecular Formula C12H20O4
CAS 220497-69-8
Enantiomeric Excess 97.5% min. (GC)
Synonym (S)-4-Cyclohexyl-3-(methoxycarbonyl)butyric acid

Specifications

Chemical Name or Material (S)-2-(Cyclohexylmethyl)succinic acid-1-methyl ester
Name Note 95%, (98% e.e.)
CAS 220497-69-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H20O4
Formula Weight 228.28
MDL Number MFCD01311762
Synonym (S)-4-Cyclohexyl-3-(methoxycarbonyl)butyric acid