Please login to your online account to display your discounted pricing

(S)-2-(Cyclohexylmethyl)succinic acid-1-methyl ester, 95%, 98% e.e., Acros Organics

$1,219.07 - $1,219.07

Chemical Identifiers

Chemical Name or Material (S)-2-(Cyclohexylmethyl)succinic acid-1-methyl ester
Name Note 95%, (98% e.e.)
CAS 220497-69-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H20O4
Formula Weight 228.28
MDL Number MFCD01311762
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362082500

  SDS
thermo fisher scientific
362082500
Glass Ampule with screw-cap 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC362080010

  SDS
thermo fisher scientific
362080010
Glass bottle 1g Each for $1,219.07
Description & Specifications

Specifications

Assay ≥94%
Physical Form Viscous Liquid
Formula Weight 228.28
Quantity 250mg
Color Clear colorless to pale yellow
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Packaging Glass Ampule with screw-cap
Assay Percent Range 95%
MDL Number MFCD01311762
Chemical Name or Material (S)-2-(Cyclohexylmethyl)succinic acid-1-methyl ester
Molecular Formula C12H20O4
CAS 220497-69-8
Name Note 95%, (98% e.e.)
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (GC)
Synonym (S)-4-Cyclohexyl-3-(methoxycarbonyl)butyric acid