Learn More
(S)-(-)-2-Chloropropionic acid, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC254970010
Specifications
(S)-(-)-2-Chloropropionic acid | |
107°C | |
Glass bottle | |
1.24 | |
-14.5 | |
Yellow | |
1g | |
29617-66-1 | |
100.0 | |
C3H5ClO2 | |
2 s-chloropropionic acid, 2s-2-chloropropanoic acid, 2s-chloropropanoic acid, alpha-l-chloropropionic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, unii-pcc7j322cm | |
GAWAYYRQGQZKCR-REOHCLBHSA-N | |
(2S)-2-chloropropanoic acid | |
107915 | |
108.52 | |
98% |
1.2400g/mL | |
Authentic | |
1.4340 to 1.436 | |
− 14.50 (20.00°C c=neat) | |
187°C | |
4°C | |
98% | |
97.5 | |
97.5% min. (GC) | |
MFCD00064205 | |
Solubility in water: soluble in water. Other solubilities: soluble in ethanol and ether | |
CC(C(=O)O)Cl | |
108.52 | |
CHEBI:73956 | |
Liquid |
Chemical Identifiers
29617-66-1 | |
108.52 | |
GAWAYYRQGQZKCR-REOHCLBHSA-N | |
107915 | |
(2S)-2-chloropropanoic acid |
C3H5ClO2 | |
MFCD00064205 | |
2 s-chloropropionic acid, 2s-2-chloropropanoic acid, 2s-chloropropanoic acid, alpha-l-chloropropionic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, unii-pcc7j322cm | |
CHEBI:73956 | |
CC(C(=O)O)Cl |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RTECSNumber : UA2451950
TSCA : TSCA