Please login to your online account to display your discounted pricing

(S)-(+)-2-chloromandelic Acid 99%, ACROS Organics™

$110.30 - $110.30

Chemical Identifiers

Chemical Name or Material (S)-(+)-2-Chloromandelic acid
CAS 52950-19-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8 H7 Cl O3
Linear Formula ClC6H4CH2(OH)COOH
Formula Weight 186.59
MDL Number MFCD00798437
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC382870010

  SDS
thermo fisher scientific
AC382870010
Glass bottle 1g Each for $110.30

AC382872500

  SDS
thermo fisher scientific
AC382872500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 186.59
Physical Form Powder
Quantity 1g
Color White to light brown
Melting Point 118° to 122°C
CAS Max % 100.0
Linear Formula ClC6H4CH2(OH)COOH
Infrared Spectrum Authentic
Molecular Formula C8 H7 Cl O3
Chemical Name or Material (S)-(+)-2-Chloromandelic acid
MDL Number MFCD00798437
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
CAS Min % 98.5
Packaging Glass bottle
CAS 52950-19-3
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.