Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-2-Chloromandelic acid, 95%

Chemical Identifiers

Chemical Name or Material (S)-(+)-2-Chloromandelic acid
CAS 52950-19-3
Formula Weight 186.59
MDL Number MFCD00798437
Synonym (S)-2-(2-Chlorophenyl)-2-hydroxyacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6115203

 
alfa aesar™
H6115203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6115206

 
alfa aesar™
H6115206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6115214

 
alfa aesar™
H6115214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.59
Melting Point 118° to 122°C
Chemical Name or Material (S)-(+)-2-Chloromandelic acid
Physical Form Powder
Assay 95%
MDL Number MFCD00798437
CAS 52950-19-3
Synonym (S)-2-(2-Chlorophenyl)-2-hydroxyacetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White to pale brown
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.