Please login to your online account to display your discounted pricing

(S)-(-)-2-(Carbobenzyloxyamino)-3-phenyl-1-propanol, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  334710010

 View more versions of this product

Catalog No. AC334710010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation -41° to -45° (20°C, 589nm) (c=2, MeOH)
Color White
Melting Point 90° to 94°C
Quantity 1g
Chemical Name or Material (S)-(-)-2-(Carbobenzyloxyamino)-3-phenyl-1-propanol
Name Note 98%
CAS 6372-14-1
CAS Min % 97.5
CAS Max % 100.0
Physical Form White powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C17H19NO3
Formula Weight 285.34g/mol
MDL Number MFCD00191138
TSCA TSCA

Specifications

Chemical Name or Material (S)-(-)-2-(Carbobenzyloxyamino)-3-phenyl-1-propanol
Name Note 98%
CAS 6372-14-1
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C17H19NO3
Formula Weight 285.34g/mol
MDL Number MFCD00191138