Please login to your online account to display your discounted pricing

(S)-(-)-2-(Carbobenzyloxyamino)-3-phenyl-1-propanol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-(-)-2-(Carbobenzyloxyamino)-3-phenyl-1-propanol
Name Note 98%
CAS 6372-14-1
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C17H19NO3
Formula Weight 285.34g/mol
MDL Number MFCD00191138
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334710010

  SDS
thermo fisher scientific
334710010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation -41° to -45° (20°C, 589nm) (c=2, MeOH)
Color White
Melting Point 90° to 94°C
Quantity 1g
Chemical Name or Material (S)-(-)-2-(Carbobenzyloxyamino)-3-phenyl-1-propanol
Name Note 98%
CAS 6372-14-1
CAS Min % 97.5
CAS Max % 100.0
Physical Form White powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C17H19NO3
Formula Weight 285.34g/mol
MDL Number MFCD00191138
TSCA TSCA