missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(-)-2-Bromopropionic acid, 98%, >85% ee, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 277910050
Specifications
(S)-(-)-2-Bromopropionic acid | |
80% min. (Typically) | |
Authentic | |
1.4740 to 1.476 | |
-27 | |
Colorless | |
5g | |
32644-15-8 | |
100.0 | |
C3H5BrO2 | |
MFCD00137325 | |
2s---2-bromopropionic acid, 2s-2-bromopropanoic acid, l-2-bromopropionic acid, l-alpha-bromopropionic acid, s---2-bromopropionic acid, s---2-bromopropionicacid, s-2-bromopropanoic acid, s-2-bromopropionic acid, s-alpha-bromopropionic acid | |
MONMFXREYOKQTI-REOHCLBHSA-N | |
(2S)-2-bromopropanoic acid | |
642232 | |
Liquid |
1.7000g/mL | |
100°C | |
Glass bottle | |
1.7 | |
203°C | |
-35°C | |
>85%ee | |
97.5 | |
97.5% min. (HPLC) | |
CH3CH(Br)CO2H | |
02, 254 | |
Solubility in water: soluble. Other solubilities: soluble in organic solvents | |
CC(C(=O)O)Br | |
152.98 | |
152.98 | |
98%,>85% ee |
Chemical Identifiers
32644-15-8 | |
152.98 | |
MONMFXREYOKQTI-REOHCLBHSA-N | |
642232 | |
CC(C(=O)O)Br |
C3H5BrO2 | |
MFCD00137325 | |
2s---2-bromopropionic acid, 2s-2-bromopropanoic acid, l-2-bromopropionic acid, l-alpha-bromopropionic acid, s---2-bromopropionic acid, s---2-bromopropionicacid, s-2-bromopropanoic acid, s-2-bromopropionic acid, s-alpha-bromopropionic acid | |
(2S)-2-bromopropanoic acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger