missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-2-(Boc-aminomethyl)-4,4-diethoxypiperidine fumarate

$105.30 - $105.30

Chemical Identifiers

Molecular Formula C15H30N2O4·C4H4O4
Formula Weight 418.49
MDL Number MFCD09953464
Synonym (S)-1-(tert-Butoxycarbonyl)-4,4-diethoxypiperidine-2-methylamine
Chemical Name or Material (S)-(-)-2-(Boc-aminomethyl)-4,4-diethoxypiperidine fumarate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH25907MC SDS Alfa Aesar™
H25907MC
100mg Each for $105.30
AAH25907ME SDS Alfa Aesar™
H25907ME
500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

C15H30N2O4·C4H4O4
MFCD09953464
(S)-(-)-2-(Boc-aminomethyl)-4,4-diethoxypiperidine fumarate
418.49
(S)-1-(tert-Butoxycarbonyl)-4,4-diethoxypiperidine-2-methylamine

Specifications

C15H30N2O4·C4H4O4
MFCD09953464
100mg
418.49
(S)-1-(tert-Butoxycarbonyl)-4,4-diethoxypiperidine-2-methylamine
(S)-(-)-2-(Boc-aminomethyl)-4,4-diethoxypiperidine fumarate

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning