Please login to your online account to display your discounted pricing

(S)-2-(BOC-amino)-1-propanol, 98% , ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC425980050

 View more versions of this product

Catalog No. AC425980050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 175.23
Physical Form Crystalline Powder or Lumps
Quantity 5g
Color White
Melting Point 59° to 62°C
CAS Max % 100.0
Molecular Formula C8H17NO3
CAS 79069-13-9
NMR Consistent with structure
Linear Formula HOCH2CH(CH3)NHCO2C(CH3)3
Chemical Name or Material (S)-2-(BOC-Amino)-1-propanol
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00043121
CAS Min % 97.5
Specific Rotation -9.5° to -11.5° (20°C, 589nm) (c=1, CHCl3)
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (S)-2-(BOC-Amino)-1-propanol
CAS 79069-13-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H17NO3
Linear Formula HOCH2CH(CH3)NHCO2C(CH3)3
Formula Weight 175.23
MDL Number MFCD00043121
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.