Please login to your online account to display your discounted pricing

(S)-2-(BOC-amino)-1-propanol, 98% , ACROS Organics™

$166.25 - $166.25

Chemical Identifiers

Chemical Name or Material (S)-2-(BOC-Amino)-1-propanol
CAS 79069-13-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H17NO3
Linear Formula HOCH2CH(CH3)NHCO2C(CH3)3
Formula Weight 175.23
MDL Number MFCD00043121
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC425980010

 
thermo fisher scientific
AC425980010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC425980050

 
thermo fisher scientific
AC425980050
Glass bottle 5g Each for $166.25
Description & Specifications

Specifications

Physical Form Crystalline Powder or Lumps
Assay ≥97.5%
Formula Weight 175.23
Quantity 1g
Color White
Melting Point 59° to 62°C
CAS Max % 100.0
Chemical Name or Material (S)-2-(BOC-Amino)-1-propanol
NMR Consistent with structure
Specific Rotation -9.5° to -11.5° (20°C, 589nm) (c=1, CHCl3)
Linear Formula HOCH2CH(CH3)NHCO2C(CH3)3
Assay Percent Range 98%
Molecular Formula C8H17NO3
MDL Number MFCD00043121
CAS 79069-13-9
Packaging Glass bottle
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.