missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-2-Benzylsuccinic acid 1-methyl ester, 98+%, ee 98+%

$274.50 - $789.57

Chemical Identifiers

CAS 182247-45-6
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD01311761
InChI Key BUNMUVFKMIOEQU-JTQLQIEISA-N
Synonym s-3-benzyl-4-methoxy-4-oxobutanoic acid, s---2-benzylsuccinic acid 1-methyl ester, 3s-3-benzyl-4-methoxy-4-oxobutanoic acid, s-2-benzylsuccinic acid 1-methyl ester, 3s-3-methoxycarbonyl-4-phenylbutanoic acid, butanedioic acid, phenylmethyl-, 1-methyl ester, 2s-9ci
PubChem CID 7006556
IUPAC Name (3S)-3-benzyl-4-methoxy-4-oxobutanoic acid
SMILES COC(=O)C(CC1=CC=CC=C1)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1826503 View Documents Alfa Aesar™
L1826503
1g Each for $789.57
 
AAL18265MD View Documents Alfa Aesar™
L18265MD
250mg Each for $274.50
 
Specifications

Chemical Identifiers

182247-45-6
222.24
BUNMUVFKMIOEQU-JTQLQIEISA-N
7006556
COC(=O)C(CC1=CC=CC=C1)CC(=O)O
C12H14O4
MFCD01311761
s-3-benzyl-4-methoxy-4-oxobutanoic acid, s---2-benzylsuccinic acid 1-methyl ester, 3s-3-benzyl-4-methoxy-4-oxobutanoic acid, s-2-benzylsuccinic acid 1-methyl ester, 3s-3-methoxycarbonyl-4-phenylbutanoic acid, butanedioic acid, phenylmethyl-, 1-methyl ester, 2s-9ci
(3S)-3-benzyl-4-methoxy-4-oxobutanoic acid

Specifications

1g
C12H14O4
7638790
BUNMUVFKMIOEQU-JTQLQIEISA-N
(3S)-3-benzyl-4-methoxy-4-oxobutanoic acid
7006556
≥98%
ee ≥98%
182247-45-6
MFCD01311761
s-3-benzyl-4-methoxy-4-oxobutanoic acid, s---2-benzylsuccinic acid 1-methyl ester, 3s-3-benzyl-4-methoxy-4-oxobutanoic acid, s-2-benzylsuccinic acid 1-methyl ester, 3s-3-methoxycarbonyl-4-phenylbutanoic acid, butanedioic acid, phenylmethyl-, 1-methyl ester, 2s-9ci
COC(=O)C(CC1=CC=CC=C1)CC(=O)O
222.24
222.24
(S)-(-)-2-Benzylsuccinic acid 1-methyl ester