missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-2-Benzylsuccinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 362072500

 View more versions of this product

Catalog No. AC362072500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
Viscous Liquid
Yellow
94.0
(S)-3-Methoxycarbonyl-4-phenylbutyric acid
(S)-2-Benzylsuccinic acid-1-methyl ester
182247-45-6
95%
Authentic
222.24
250mg
Glass Ampule with Screw-cap
100.0
95%, (98% e.e.)
MFCD01311761
C12 H14 O4
97.5% min. (HPLC)

Chemical Identifiers

182247-45-6
100.0
C12 H14 O4
MFCD01311761
(S)-2-Benzylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
94.0
95%
222.24
(S)-3-Methoxycarbonyl-4-phenylbutyric acid
≥94%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA