missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-2-Benzylsuccinic acid-1-methyl ester, 95%, 98% ee, ACROS Organics™

Chemical Identifiers

CAS 182247-45-6
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD01311761
InChI Key BUNMUVFKMIOEQU-JTQLQIEISA-N
Synonym 3s-3-benzyl-4-methoxy-4-oxobutanoic acid, 3s-3-methoxycarbonyl-4-phenylbutanoic acid, butanedioic acid, phenylmethyl-, 1-methyl ester, 2s-9ci, s---2-benzylsuccinic acid 1-methyl ester, s-2-benzylsuccinic acid 1-methyl ester, s-3-benzyl-4-methoxy-4-oxobutanoic acid
PubChem CID 7006556
IUPAC Name (3S)-3-benzyl-4-methoxy-4-oxobutanoic acid
SMILES COC(=O)C(CC1=CC=CC=C1)CC(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC362072500
SDS View Documents
Thermo Fisher Scientific
362072500
250mg Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

182247-45-6
222.24
BUNMUVFKMIOEQU-JTQLQIEISA-N
7006556
COC(=O)C(CC1=CC=CC=C1)CC(=O)O
C12H14O4
MFCD01311761
3s-3-benzyl-4-methoxy-4-oxobutanoic acid, 3s-3-methoxycarbonyl-4-phenylbutanoic acid, butanedioic acid, phenylmethyl-, 1-methyl ester, 2s-9ci, s---2-benzylsuccinic acid 1-methyl ester, s-2-benzylsuccinic acid 1-methyl ester, s-3-benzyl-4-methoxy-4-oxobutanoic acid
(3S)-3-benzyl-4-methoxy-4-oxobutanoic acid

Specifications

97.5% min. (HPLC)
Glass Bottle
250mg
94.0
94% min. (GC)
MFCD01311761
BUNMUVFKMIOEQU-JTQLQIEISA-N
(3S)-3-benzyl-4-methoxy-4-oxobutanoic acid
7006556
Viscous Liquid
(S)-2-Benzylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
Authentic
Yellow
182247-45-6
100.0
C12H14O4
3s-3-benzyl-4-methoxy-4-oxobutanoic acid, 3s-3-methoxycarbonyl-4-phenylbutanoic acid, butanedioic acid, phenylmethyl-, 1-methyl ester, 2s-9ci, s---2-benzylsuccinic acid 1-methyl ester, s-2-benzylsuccinic acid 1-methyl ester, s-3-benzyl-4-methoxy-4-oxobutanoic acid
COC(=O)C(CC1=CC=CC=C1)CC(=O)O
222.24
222.24
95%,98% ee
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS