missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-2-(Benzyloxycarbonylamino)butyric acid, 98%

$53.05 - $784.93

Chemical Identifiers

CAS 42918-86-5
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00077006
InChI Key SZQMTCSQWUYUML-JTQLQIEISA-N
Synonym z-abu-oh, s-2-benzyloxy carbonyl amino butanoic acid, 2s-2-benzyloxy carbonyl amino butanoic acid, z-2-abu-oh, s-2-bezyloxycarbonylamino butanoic acid, s-2-benzyloxycarbonylamino butanoic acid, butanoic acid, 2-phenylmethoxy carbonyl amino-, 2s, ambotzzaa1200, pubchem18943, s-2-cbz-amino butyricacid
PubChem CID 7349998
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6242603 View Documents Alfa Aesar™
H6242603
1g Each for $53.05
 
AAH6242606 View Documents Alfa Aesar™
H6242606
5g Each for $195.85
 
AAH6242614 View Documents Alfa Aesar™
H6242614
25g Each for $784.93
 
Specifications

Chemical Identifiers

42918-86-5
237.255
SZQMTCSQWUYUML-JTQLQIEISA-N
7349998
CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
C12H15NO4
MFCD00077006
z-abu-oh, s-2-benzyloxy carbonyl amino butanoic acid, 2s-2-benzyloxy carbonyl amino butanoic acid, z-2-abu-oh, s-2-bezyloxycarbonylamino butanoic acid, s-2-benzyloxycarbonylamino butanoic acid, butanoic acid, 2-phenylmethoxy carbonyl amino-, 2s, ambotzzaa1200, pubchem18943, s-2-cbz-amino butyricacid
(2S)-2-(phenylmethoxycarbonylamino)butanoic acid

Specifications

1g
C12H15NO4
z-abu-oh, s-2-benzyloxy carbonyl amino butanoic acid, 2s-2-benzyloxy carbonyl amino butanoic acid, z-2-abu-oh, s-2-bezyloxycarbonylamino butanoic acid, s-2-benzyloxycarbonylamino butanoic acid, butanoic acid, 2-phenylmethoxy carbonyl amino-, 2s, ambotzzaa1200, pubchem18943, s-2-cbz-amino butyricacid
CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
237.255
237.25
(S)-2-(Benzyloxycarbonylamino)butyric acid
42918-86-5
MFCD00077006
SZQMTCSQWUYUML-JTQLQIEISA-N
(2S)-2-(phenylmethoxycarbonylamino)butanoic acid
7349998
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning