missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6277203
Description
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Store in freezer (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt | |
White | |
MFCD00237341 | |
dicyclohexylamine s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, z-dab boc-oh dcha, z-dab boc-oh?cha, z-dab boc-oh.dcha, n-a-z-n-?-boc-l-2,4-diaminobutyric aciddicyclohexylaminesalt, s-2-benzyloxycarbonylamino-4-boc-amino butyric acid dicyclohexylammonium salt, z-dab boc-oh.dcha na-z-n4-boc-2,4-diaminobutyric acid dicyclohexylamine salt, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid-n-cyclohexylcyclohexanamine 1/1, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid; dicha | |
CYMIEBREXUYHGN-UHFFFAOYNA-N | |
N-cyclohexylcyclohexanamine;(2S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)butanoic acid | |
49853427 | |
98% |
3350-13-8 | |
C29H47N3O6 | |
1 g | |
Slightly soluble in water. | |
C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O | |
533.71 | |
553.7 | |
Powder |
Chemical Identifiers
3350-13-8 | |
533.71 | |
CYMIEBREXUYHGN-UHFFFAOYNA-N | |
49853427 | |
C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O |
C29H47N3O6 | |
MFCD00237341 | |
dicyclohexylamine s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, z-dab boc-oh dcha, z-dab boc-oh?cha, z-dab boc-oh.dcha, n-a-z-n-?-boc-l-2,4-diaminobutyric aciddicyclohexylaminesalt, s-2-benzyloxycarbonylamino-4-boc-amino butyric acid dicyclohexylammonium salt, z-dab boc-oh.dcha na-z-n4-boc-2,4-diaminobutyric acid dicyclohexylamine salt, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid-n-cyclohexylcyclohexanamine 1/1, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid; dicha | |
N-cyclohexylcyclohexanamine;(2S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)butanoic acid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only