missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%

Manufacturer:  Alfa Aesar™ H62772MD

 View more versions of this product

Catalog No. AAH62772MD



Description

Description

Applications
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in freezer (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt
250mg
C29H47N3O6
dicyclohexylamine s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, z-dab boc-oh dcha, z-dab boc-oh?cha, z-dab boc-oh.dcha, n-a-z-n-?-boc-l-2,4-diaminobutyric aciddicyclohexylaminesalt, s-2-benzyloxycarbonylamino-4-boc-amino butyric acid dicyclohexylammonium salt, dicyclohexylamine s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, z-dab boc-oh.dcha na-z-n4-boc-2,4-diaminobutyric acid dicyclohexylamine salt, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid-n-cyclohexylcyclohexanamine 1/1, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid; dicha
CC(C)(C)OC(=O)NCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1.C1CCC(CC1)NC2CCCCC2
533.71
553.7
98%
White
3350-13-8
MFCD00237341
CYMIEBREXUYHGN-ZOWNYOTGSA-N
N-cyclohexylcyclohexanamine;(2S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)butanoic acid
49853427
Powder

Chemical Identifiers

3350-13-8
533.71
CYMIEBREXUYHGN-ZOWNYOTGSA-N
49853427
CC(C)(C)OC(=O)NCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1.C1CCC(CC1)NC2CCCCC2
C29H47N3O6
MFCD00237341
dicyclohexylamine s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, z-dab boc-oh dcha, z-dab boc-oh?cha, z-dab boc-oh.dcha, n-a-z-n-?-boc-l-2,4-diaminobutyric aciddicyclohexylaminesalt, s-2-benzyloxycarbonylamino-4-boc-amino butyric acid dicyclohexylammonium salt, dicyclohexylamine s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, z-dab boc-oh.dcha na-z-n4-boc-2,4-diaminobutyric acid dicyclohexylamine salt, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid-n-cyclohexylcyclohexanamine 1/1, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid; dicha
N-cyclohexylcyclohexanamine;(2S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)butanoic acid
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents