missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-2-Azetidinecarboxylic acid, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC105140050

 View more versions of this product

Catalog No. AC105140050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Crystalline Powder or Crystals
≥99.0%
White to light beige
99.0
-123° to -127° (25°C, 589nm) (c=1, water)
(S)-(-)-2-Azetidinecarboxylic acid
C4H7NO2
Authentic
101.1
5g
−125° (25°C c=1,water)
100.0
99+%
2133-34-8
Glass bottle

Chemical Identifiers

2133-34-8
100.0
C4H7NO2
(S)-(-)-2-Azetidinecarboxylic acid
99.0
99+%
101.1
≥99.0%