Please login to your online account to display your discounted pricing

(S)-(-)-2-Azetidinecarboxylic acid, 99+%, ACROS Organics™


Quantity 50mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  105140500

 View more versions of this product

Catalog No. AC105140500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Powder or Crystals
Formula Weight 101.1
Quantity 50mg
Color White to light beige
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material (S)-(-)-2-Azetidinecarboxylic acid
Infrared Spectrum Authentic
Assay Percent Range 99+%
Molecular Formula C4H7NO2
Packaging Glass bottle
CAS 2133-34-8
Specific Rotation -123° to -127° (25°C, 589nm) (c=1, water)

Specifications

Chemical Name or Material (S)-(-)-2-Azetidinecarboxylic acid
CAS 2133-34-8
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C4H7NO2
Formula Weight 101.1