Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-2-Aminooctane, ChiPros™ 99+%, ee 99+%

Chemical Identifiers

Chemical Name or Material (S)-(+)-2-Aminooctane
CAS 34566-04-6
Molecular Formula C8H19N
Formula Weight 129.25
MDL Number MFCD01091021
UN Number UN2734
Beilstein 1719319
Synonym (S)-(+)-1-Methylheptylamine; (2S)-Octylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1930903

 
alfa aesar™
L1930903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1930906

 
alfa aesar™
L1930906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1930914

 
alfa aesar™
L1930914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 129.25
MDL Number MFCD01091021
CAS 34566-04-6
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point <−20°C
Quantity 1g
Chemical Name or Material (S)-(+)-2-Aminooctane
Assay ≥99%
Molecular Formula C8H19N
Synonym (S)-(+)-1-Methylheptylamine; (2S)-Octylamine
Assay Percent Notes ee ≥99%
UN Number UN2734
Beilstein 1719319
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H314-H318-H226
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Boiling Point 163° to 165°C
Flash Point 50°C (122°F)
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H314-H318-H226
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.