Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-2-Aminohexane, ChiPros 99+%, ee 99+%

Chemical Identifiers

Chemical Name or Material (S)-(+)-2-Aminohexane
CAS 70492-67-0
Molecular Formula C6H15N
Formula Weight 101.19
MDL Number MFCD00671626
UN Number UN2733
Beilstein 5237601
Synonym (2S)-Hexylamine; (S)-1-Methylpentylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1930503

 
alfa aesar™
L1930503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1930506

 
alfa aesar™
L1930506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1930514

 
alfa aesar™
L1930514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 101.19
CAS 70492-67-0
Chemical Name or Material (S)-(+)-2-Aminohexane
MDL Number MFCD00671626
Synonym (2S)-Hexylamine; (S)-1-Methylpentylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314-H318-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P309-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Boiling Point 116°C
Melting Point <−20°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Assay Percent Notes ee ≥99%
UN Number UN2733
Beilstein 5237601
Quantity 1g
Flash Point −3°C (26°F)
Molecular Formula C6H15N
Assay ≥99%

Applications
(S)-(+)-2-Aminohexane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water (17g/L at 20°C)

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.

Safety and Handling

Danger

GHS P Statement
P210-P280-P305+P351+P338-P309-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H225-H314-H318-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.