Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-2-Aminoheptane, ChiPros 99%, ee 99+%

Chemical Identifiers

Chemical Name or Material (S)-(+)-2-Aminoheptane
CAS 44745-29-1
Molecular Formula C7H17N
Formula Weight 115.22
MDL Number MFCD00066324
UN Number UN2733
Beilstein 1719086
Synonym (2S)-(+)-Heptylamine; (S)-1-Methylhexylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1930703

 
alfa aesar™
L1930703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1930706

 
alfa aesar™
L1930706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1930714

 
alfa aesar™
L1930714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 115.22
Chemical Name or Material (S)-(+)-2-Aminoheptane
Assay Percent Notes ee ≥99%
Molecular Formula C7H17N
MDL Number MFCD00066324
CAS 44745-29-1
UN Number UN2733
Beilstein 1719086
Synonym (2S)-(+)-Heptylamine; (S)-1-Methylhexylamine
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Flash Point 54°C (129°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Assay 99%
Health Hazard 2 GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Boiling Point 142° to 144°C
Quantity 1g
Optical Rotation +7.2° (Neat)

Applications
(S)-(+)-2-Aminoheptane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water (5g/L at 20°C)

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.