missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(+)-2-Amino-4-phosphonobutyric acid, 97%, Thermo Scientific Chemicals
A NMDA agonist
$187.49 - $426.11
Chemical Identifiers
CAS | 23052-81-5 |
---|---|
Molecular Formula | C4H10NO5P |
Molecular Weight (g/mol) | 183.1 |
MDL Number | MFCD00083244 |
InChI Key | DDOQBQRIEWHWBT-VKHMYHEASA-N |
Synonym | l-ap4, 2s-2-amino-4-phosphonobutanoic acid, l-+-2-amino-4-phosphonobutyric acid, chembl33567, s-2-amino-4-phosphonobutyrate, l-ap-4, 3h ap4, l-2-amino-4-phosphonobutiric acid, l-1-amino-4-phosphonobutanoic acid, s-2-amino-4-phosphonobutanoic acid |
PubChem CID | 179394 |
IUPAC Name | (2S)-2-amino-4-phosphonobutanoic acid |
SMILES | C(CP(=O)(O)O)C(C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ64075MCR
|
Thermo Scientific Chemicals
J64075MCR |
1 mg |
Each for $187.49
|
|
|||||
AAJ64075MA
|
Thermo Scientific Chemicals
J64075MA |
10 mg |
Each for $426.11
|
|
|||||
Description
A NMDA agonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
23052-81-5 | |
183.1 | |
DDOQBQRIEWHWBT-VKHMYHEASA-N | |
179394 | |
C(CP(=O)(O)O)C(C(=O)O)N |
C4H10NO5P | |
MFCD00083244 | |
l-ap4, 2s-2-amino-4-phosphonobutanoic acid, l-+-2-amino-4-phosphonobutyric acid, chembl33567, s-2-amino-4-phosphonobutyrate, l-ap-4, 3h ap4, l-2-amino-4-phosphonobutiric acid, l-1-amino-4-phosphonobutanoic acid, s-2-amino-4-phosphonobutanoic acid | |
(2S)-2-amino-4-phosphonobutanoic acid |
Specifications
23052-81-5 | |
C4H10NO5P | |
1 mg | |
l-ap4, 2s-2-amino-4-phosphonobutanoic acid, l-+-2-amino-4-phosphonobutyric acid, chembl33567, s-2-amino-4-phosphonobutyrate, l-ap-4, 3h ap4, l-2-amino-4-phosphonobutiric acid, l-1-amino-4-phosphonobutanoic acid, s-2-amino-4-phosphonobutanoic acid | |
DDOQBQRIEWHWBT-VKHMYHEASA-N | |
C(CP(=O)(O)O)C(C(=O)O)N | |
183.1 | |
183.1 | |
Powder |
White | |
MFCD00083244 | |
Light sensitive | |
Soluble to 5 mM in water and to 100 mM in 1eq. NaOH | |
+24° (c=1 in 6N HCl) | |
(2S)-2-amino-4-phosphonobutanoic acid | |
179394 | |
99% | |
(S)-(+)-2-Amino-4-phosphonobutyric acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : EK7713874
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only