Learn More
(S)-(+)-2-Amino-3-methyl-1-butanol, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 181590050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(S)-(+)-2-Amino-3-methyl-1-butanol | |
95.0 | |
30.0°C to 32.0°C | |
0.9200g/mL | |
91°C | |
95% min. (GC) | |
C5H13NO | |
(CH3)2CHCH(NH2)CH2OH | |
5 mL | |
0.92 | |
+ 16.00 | |
NWYYWIJOWOLJNR-RXMQYKEDSA-N | |
(2S)-2-amino-3-methylbutan-1-ol | |
640993 | |
96% |
2026-48-4 | |
100.0 | |
Yellow | |
81.0°C (8.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4538 to 1.4558 | |
MFCD00064296 | |
16,38 | |
+16.00 (20.00°C c=10,ethanol) | |
l-valinol, s-+-2-amino-3-methyl-1-butanol, 2s-2-amino-3-methylbutan-1-ol, l-+-valinol, h-valinol, s-2-amino-3-methylbutan-1-ol, s-2-amino-3-methyl-1-butanol, 1-butanol, 2-amino-3-methyl-, 2s, s-2-amino-3-methyl-butan-1-ol | |
CC(C)[C@H](N)CO | |
103.17 | |
103.16 | |
Liquid After Melting |
Chemical Identifiers
2026-48-4 | |
103.17 | |
NWYYWIJOWOLJNR-RXMQYKEDSA-N | |
640993 | |
CC(C)[C@H](N)CO |
C5H13NO | |
MFCD00064296 | |
l-valinol, s-+-2-amino-3-methyl-1-butanol, 2s-2-amino-3-methylbutan-1-ol, l-+-valinol, h-valinol, s-2-amino-3-methylbutan-1-ol, s-2-amino-3-methyl-1-butanol, 1-butanol, 2-amino-3-methyl-, 2s, s-2-amino-3-methyl-butan-1-ol | |
(2S)-2-amino-3-methylbutan-1-ol |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 217-975-5
RUO – Research Use Only