Please login to your online account to display your discounted pricing

(S)-(+)-2-Amino-3-hydroxy-3-methylbutanoicacid 98% (94% e.e.), ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: fisher scientific acros  299142500

 View more versions of this product

Catalog No. AC299142500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (TLC)
Formula Weight 133.15
Physical Form Crystalline Powder
Quantity 250mg
Color White
Specific Rotation Condition − 11.75 (25.00°C c=2,6N HCl)
CAS 2280-27-5
Assay Percent Range 98%
Linear Formula CH3C(CH3)(OH)CH(NH2)COOH
Infrared Spectrum Authentic
Enantiomeric Excess 93% min. (HPLC)
Synonym L(+)-Hydroxyvaline
Packaging Glass bottle
Specific Rotation +11° to +14° (20°C, 589nm) (c=2, 6 N HCl)
Individual Impurity 97.5% min. (TLC)
Chemical Name or Material (S)-(+)-2-Amino-3-hydroxy-3-methylbutanoic acid
Molecular Formula C5H11NO3

Specifications

Chemical Name or Material (S)-(+)-2-Amino-3-hydroxy-3-methylbutanoic acid
Assay ≥97.5% (TLC)
Assay Percent Range 98%
Molecular Formula C5H11NO3
Linear Formula CH3C(CH3)(OH)CH(NH2)COOH
Formula Weight 133.15
Synonym L(+)-Hydroxyvaline