Learn More
(S)-(+)-2-Amino-1-phenylethanol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1940406
Description
(S)-(+)-2-Amino-1-phenylethanol is used as an intermediate in organic synthesis. It is involved in the preparation of enantiopure (R)-octopamine, (R)-tembamide and (R)-aegeline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(+)-2-Amino-1-phenylethanol is used as an intermediate in organic synthesis. It is involved in the preparation of enantiopure (R)-octopamine, (R)-tembamide and (R)-aegeline.
Solubility
Soluble in methanol.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
Specifications
(S)-(+)-2-Amino-1-phenylethanol | |
57°C to 63°C | |
C8H11NO | |
5 g | |
2412760 | |
s-2-amino-1-phenylethanol, s---2-phenylglycinol, 1s-2-amino-1-phenylethanol, s-2-amino-1-phenylethan-1-ol, 1s-2-amino-1-phenylethan-1-ol, s-+-2-amino-1-phenylethanol, r---alpha-aminomethyl benzyl alcohol, pubchem13836, 2-amino-1 s-phenylethanol | |
ULSIYEODSMZIPX-MRVPVSSYSA-N | |
NC[C@@H](O)C1=CC=CC=C1 | |
137.18 | |
137.18 |
56613-81-1 | |
160°C (17 mmHg) | |
MFCD00239405 | |
UN3259 | |
Air Sensitive | |
Soluble in methanol. | |
+42° (c=2 in Ethanol) | |
(1S)-2-amino-1-phenylethanol | |
643217 | |
97% |
Chemical Identifiers
56613-81-1 | |
137.18 | |
ULSIYEODSMZIPX-MRVPVSSYSA-N | |
643217 | |
NC[C@@H](O)C1=CC=CC=C1 |
C8H11NO | |
MFCD00239405 | |
s-2-amino-1-phenylethanol, s---2-phenylglycinol, 1s-2-amino-1-phenylethanol, s-2-amino-1-phenylethan-1-ol, 1s-2-amino-1-phenylethan-1-ol, s-+-2-amino-1-phenylethanol, r---alpha-aminomethyl benzyl alcohol, pubchem13836, 2-amino-1 s-phenylethanol | |
(1S)-2-amino-1-phenylethanol |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only