missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-2-Amino-1-diphenylphosphino-3-methylbutane, 97+%

$133.70 - $450.30

Chemical Identifiers

CAS 146476-37-1
Molecular Formula C17H22NP
Molecular Weight (g/mol) 271.344
MDL Number MFCD17013993
InChI Key ZZLCXURCZWQECA-QGZVFWFLSA-N
Synonym s-1-diphenylphosphino-3-methylbutan-2-amine, s-1-diphenylphosphino-2-amino-3-methylbutane, 2s-1-diphenylphosphanyl-3-methylbutan-2-amine, s-2-amino-1-diphenylphosphino-3-methylbutane, 2s-2-amino-3-methylbutyl diphenylphosphane, 2s-2-amino-3-methylbutyl diphenylphosphine, s-1-diphenylphosphino-3-methyl-2-butylamine, s-1-diphenylphosphino-3-methyl-2-butylamine, s-1-diphenylphosphino-2-amino-3-methylbutane,min
PubChem CID 15145880
IUPAC Name (2S)-1-diphenylphosphanyl-3-methylbutan-2-amine
SMILES CC(C)C(CP(C1=CC=CC=C1)C2=CC=CC=C2)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60980MD View Documents Alfa Aesar™
H60980MD
250mg Each for $133.70
 
AAH6098003 View Documents Alfa Aesar™
H6098003
1g Each for $450.30
 
Specifications

Chemical Identifiers

146476-37-1
271.344
ZZLCXURCZWQECA-QGZVFWFLSA-N
15145880
CC(C)C(CP(C1=CC=CC=C1)C2=CC=CC=C2)N
C17H22NP
MFCD17013993
s-1-diphenylphosphino-3-methylbutan-2-amine, s-1-diphenylphosphino-2-amino-3-methylbutane, 2s-1-diphenylphosphanyl-3-methylbutan-2-amine, s-2-amino-1-diphenylphosphino-3-methylbutane, 2s-2-amino-3-methylbutyl diphenylphosphane, 2s-2-amino-3-methylbutyl diphenylphosphine, s-1-diphenylphosphino-3-methyl-2-butylamine, s-1-diphenylphosphino-3-methyl-2-butylamine, s-1-diphenylphosphino-2-amino-3-methylbutane,min
(2S)-1-diphenylphosphanyl-3-methylbutan-2-amine

Specifications

>110°C (230°F)
146476-37-1
MFCD17013993
s-1-diphenylphosphino-3-methylbutan-2-amine, s-1-diphenylphosphino-2-amino-3-methylbutane, 2s-1-diphenylphosphanyl-3-methylbutan-2-amine, s-2-amino-1-diphenylphosphino-3-methylbutane, 2s-2-amino-3-methylbutyl diphenylphosphane, 2s-2-amino-3-methylbutyl diphenylphosphine, s-1-diphenylphosphino-3-methyl-2-butylamine, s-1-diphenylphosphino-3-methyl-2-butylamine, s-1-diphenylphosphino-2-amino-3-methylbutane,min
CC(C)C(CP(C1=CC=CC=C1)C2=CC=CC=C2)N
271.344
271.34
(S)-2-Amino-1-diphenylphosphino-3-methylbutane
250mg
C17H22NP
UN2735
ZZLCXURCZWQECA-QGZVFWFLSA-N
(2S)-1-diphenylphosphanyl-3-methylbutan-2-amine
15145880
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.