Please login to your online account to display your discounted pricing

(S)-(+)-2-Amino-1-butanol ≥98% (ca 95% e.e.), ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  158080010

 View more versions of this product

Catalog No. AC158080010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Viscous Liquid
Assay ≥98.0%
Formula Weight 89.14
Quantity 1g
Color Clear colorless to slightly yellow
EINECS Number 227-475-9
Merck Index 10, 429
Beilstein 04, 291
CAS 5856-62-2
Synonym (S)-(+)-2-Amino-1-butanol
Health Hazard 1 GHS Signal Word: Danger
Name Note 98+% (ca 95% e.e.)
Assay Percent Range 98+%
Linear Formula C2H5CH(NH2)CH2OH
Chemical Name or Material (S)-(+)-2-Amino-1-butanol
MDL Number MFCD00064418
Molecular Formula C4H11NO
Infrared Spectrum Authentic
Specific Rotation +8.5° to +11.5° (20°C, 589nm) (neat)
Packaging Glass bottle
Refractive Index 1.4515 to 1.4535 (20°C, 589nm)
Enantiomeric Excess 94% min.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (S)-(+)-2-Amino-1-butanol
Name Note 98+% (ca 95% e.e.)
CAS 5856-62-2
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C4H11NO
Linear Formula C2H5CH(NH2)CH2OH
Formula Weight 89.14
MDL Number MFCD00064418
Synonym (S)-(+)-2-Amino-1-butanol
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents