Please login to your online account to display your discounted pricing

(S)-(+)-2-Amino-1-butanol ≥98% (ca 95% e.e.), ACROS Organics™

$48.44 - $48.44

Chemical Identifiers

Chemical Name or Material (S)-(+)-2-Amino-1-butanol
Name Note 98+% (ca 95% e.e.)
CAS 5856-62-2
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C4H11NO
Linear Formula C2H5CH(NH2)CH2OH
Formula Weight 89.14
MDL Number MFCD00064418
Synonym (S)-(+)-2-Amino-1-butanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC158080010

  SDS
thermo fisher scientific
158080010
Glass bottle 1g Each for $48.44
Description & Specifications

Specifications

Physical Form Viscous Liquid
Assay ≥98.0%
Formula Weight 89.14
Quantity 1g
Color Clear colorless to slightly yellow
EINECS Number 227-475-9
Beilstein 04, 291
Merck Index 10, 429
Chemical Name or Material (S)-(+)-2-Amino-1-butanol
Name Note 98+% (ca 95% e.e.)
Assay Percent Range 98+%
Linear Formula C2H5CH(NH2)CH2OH
MDL Number MFCD00064418
CAS 5856-62-2
Synonym (S)-(+)-2-Amino-1-butanol
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Refractive Index 1.4515 to 1.4535 (20°C, 589nm)
Molecular Formula C4H11NO
Enantiomeric Excess 94% min.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Specific Rotation +8.5° to +11.5° (20°C, 589nm) (neat)