missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-2-(alpha-Methylbenzylamino)-5-nitropyridine, TCI America™

Manufacturer:  TCI America M09855G

 View more versions of this product

Catalog No. M09855G



Specifications

Specifications

(S)-(-)-2-(alpha-Methylbenzylamino)-5-nitropyridine
83°C
84249-39-8
MFCD00145247
CC(C1=CC=CC=C1)NC2=NC=C(C=C2)[N+](=O)[O-]
243.266
243.27
≥99.0% (T)
Yellow
5g
C13H13N3O2
RAKMWGKBCDCUDX-JTQLQIEISA-N
5-nitro-N-[(1S)-1-phenylethyl]pyridin-2-amine
29772754
Crystal-Powder at 20°C

Chemical Identifiers

84249-39-8
243.266
RAKMWGKBCDCUDX-JTQLQIEISA-N
5-nitro-N-[(1S)-1-phenylethyl]pyridin-2-amine
C13H13N3O2
MFCD00145247
29772754
CC(C1=CC=CC=C1)NC2=NC=C(C=C2)[N+](=O)[O-]
Safety and Handling

Safety and Handling

TSCA : No

Documents