missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-2-(alpha-Methylbenzylamino)-5-nitropyridine, TCI America™

$209.31 - $695.16

Chemical Identifiers

CAS 84249-39-8
Molecular Formula C13H13N3O2
Molecular Weight (g/mol) 243.266
MDL Number MFCD00145247
InChI Key RAKMWGKBCDCUDX-JTQLQIEISA-N
PubChem CID 29772754
IUPAC Name 5-nitro-N-[(1S)-1-phenylethyl]pyridin-2-amine
SMILES CC(C1=CC=CC=C1)NC2=NC=C(C=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M09851G View Documents TCI America
M09851G
1g Each for $209.31
 
M09855G View Documents TCI America
M09855G
5g Each for $695.16
 
Specifications

Chemical Identifiers

84249-39-8
243.266
RAKMWGKBCDCUDX-JTQLQIEISA-N
5-nitro-N-[(1S)-1-phenylethyl]pyridin-2-amine
C13H13N3O2
MFCD00145247
29772754
CC(C1=CC=CC=C1)NC2=NC=C(C=C2)[N+](=O)[O-]

Specifications

Yellow
1g
C13H13N3O2
RAKMWGKBCDCUDX-JTQLQIEISA-N
5-nitro-N-[(1S)-1-phenylethyl]pyridin-2-amine
29772754
Crystal-Powder at 20°C
(S)-(-)-2-(alpha-Methylbenzylamino)-5-nitropyridine
83°C
84249-39-8
MFCD00145247
CC(C1=CC=CC=C1)NC2=NC=C(C=C2)[N+](=O)[O-]
243.266
243.27
≥99.0% (T)
Safety and Handling

Safety and Handling

TSCA : No