Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-2-Acetoxysuccinic anhydride, 98%

$23.80 - $385.73

Chemical Identifiers

Chemical Name or Material (S)- (-)-2-Acetoxysuccinic anhydride
CAS 59025-03-5
Molecular Formula C6H6O5
Formula Weight 158.11
MDL Number MFCD00082708
Synonym L-O-Acetylmalic anhydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4436604

  SDS
alfa aesar™
4436604
2g Each for $23.80

AA4436609

  SDS
alfa aesar™
4436609
10g Each for $94.03

AA4436618

  SDS
alfa aesar™
4436618
50g Each for $385.73
Description & Specifications

Specifications

Formula Weight 158.11
Assay 98%
Sensitivity Moisture Sensitive
CAS 59025-03-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 56° to 58°C
Molecular Formula C6H6O5
Health Hazard 1 Warning
Quantity 2g
Chemical Name or Material (S)- (-)-2-Acetoxysuccinic anhydride
Synonym L-O-Acetylmalic anhydride
Physical Form Powder
MDL Number MFCD00082708
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.