missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-2-Acetoxysuccinic anhydride, 98%

Manufacturer:  Alfa Aesar™ 4436609

 View more versions of this product

Catalog No. AA4436609



Specifications

Specifications

(S)- (-)-2-Acetoxysuccinic anhydride
56°C to 58°C
59025-03-5
MFCD00082708
Reacts in water.
CC(=O)OC1CC(=O)OC1=O
158.109
158.11
98%
Moisture sensitive
10g
C6H6O5
--o-acetyl-l-malic anhydride
SSWJHSASZZAIAU-BYPYZUCNSA-N
[(3S)-2,5-dioxooxolan-3-yl] acetate
11126528
Powder

Chemical Identifiers

59025-03-5
158.109
SSWJHSASZZAIAU-BYPYZUCNSA-N
11126528
CC(=O)OC1CC(=O)OC1=O
C6H6O5
MFCD00082708
--o-acetyl-l-malic anhydride
[(3S)-2,5-dioxooxolan-3-yl] acetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents