missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-2,4-Bis(Fmoc-amino)butyric acid, 95%

$47.35 - $467.26

Chemical Identifiers

CAS 201473-83-8
Molecular Formula C34H30N2O6
Molecular Weight (g/mol) 562.622
MDL Number MFCD00237004
InChI Key ALZDIZDLDRWFAC-HKBQPEDESA-N
Synonym fmoc-dab fmoc-oh, 2s-2,4-bis 9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, butanoicacid, 2,4-bis 9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, s-2,4-bis 9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2s-2,4-bis 9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, ambotzfaa1770, fmoc-l-dab fmoc-oh, n-a,n-?-di-fmoc-l-2,4-diaminobutyric acid, n-, a,n-, a-di-fmoc-l-2,4-diaminobutyric acid, 2s-2,4-bis 9h-fluoren-9-ylmethoxycarbonylamino butanoic acid
PubChem CID 46737279
IUPAC Name (2S)-2,4-bis(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66073MD View Documents Alfa Aesar™
H66073MD
250mg Each for $47.35
 
AAH6607303 View Documents Alfa Aesar™
H6607303
1g Each for $124.91
 
AAH6607306 View Documents Alfa Aesar™
H6607306
5g Each for $467.26
 
Specifications

Chemical Identifiers

201473-83-8
562.622
ALZDIZDLDRWFAC-HKBQPEDESA-N
46737279
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
C34H30N2O6
MFCD00237004
fmoc-dab fmoc-oh, 2s-2,4-bis 9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, butanoicacid, 2,4-bis 9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, s-2,4-bis 9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2s-2,4-bis 9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, ambotzfaa1770, fmoc-l-dab fmoc-oh, n-a,n-?-di-fmoc-l-2,4-diaminobutyric acid, n-, a,n-, a-di-fmoc-l-2,4-diaminobutyric acid, 2s-2,4-bis 9h-fluoren-9-ylmethoxycarbonylamino butanoic acid
(2S)-2,4-bis(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid

Specifications

250mg
C34H30N2O6
fmoc-dab fmoc-oh, 2s-2,4-bis 9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, butanoicacid, 2,4-bis 9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, s-2,4-bis 9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2s-2,4-bis 9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, ambotzfaa1770, fmoc-l-dab fmoc-oh, n-a,n-?-di-fmoc-l-2,4-diaminobutyric acid, n-, a,n-, a-di-fmoc-l-2,4-diaminobutyric acid, 2s-2,4-bis 9h-fluoren-9-ylmethoxycarbonylamino butanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
562.622
562.62
95%
201473-83-8
MFCD00237004
ALZDIZDLDRWFAC-HKBQPEDESA-N
(2S)-2,4-bis(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
46737279
Solid
(S)-2,4-Bis(Fmoc-amino)butyric acid
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C