Learn More
(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 296470050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione | |
88.0 | |
Authentic | |
Glass bottle | |
MFCD00012272 | |
+331° (20°C c=1,toluene) | |
7as-7a-methyl-2,3,6,7-tetrahydroindene-1,5-dione, s-+-2,3,7,7a-tetrahydro-7a-methyl-1h-indene-1,5 6h-dione, 1h-indene-1,5 6h-dione, 2,3,7,7a-tetrahydro-7a-methyl-, 7as, hajos dione, s-hajos dione, s-hajos ketone, s-hajos-wiechert ketone, +-hajos-parrish ketone, +-hajos-parrish diketone, 7as-7a-methyl-2,3,7,7a-tetrahydro-1h-indene-1,5 6h-dione | |
CC12CCC(=O)C=C1CCC2=O | |
164.2 | |
164.2 |
17553-86-5 | |
100.0 | |
88% min. (GC) | |
C10H12O2 | |
5g | |
+ 331.00 | |
FNYAZSZTENLTRT-JTQLQIEISA-N | |
(7aS)-7a-methyl-2,3,6,7-tetrahydroindene-1,5-dione | |
736943 | |
90% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
GHS Signal Word: Warning
RUO â Research Use Only