missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-2,2'-Diamino-1,1'-binaphthalene, 99%, ACROS Organics™

Chemical Identifiers

CAS 18531-95-8
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C20H16N2
Formula Weight 284.36
MDL Number MFCD00145204
Chemical Name or Material (S)-(-)-2, 2'-Diamino-1, 1'-binaphthalene
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC356522500 SDS Thermo Fisher Scientific
AC356522500
Glass bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC356520010 SDS Thermo Fisher Scientific
356520010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

18531-95-8
100.0
C20H16N2
MFCD00145204
≥98.5%
98.5
99%
284.36
(S)-(-)-2, 2'-Diamino-1, 1'-binaphthalene

Specifications

−156° (20°C c=1,pyridine)
250mg
98.5
Authentic
-153° to -159° (20°C, 589nm) (c=1, pyridine)
99%
284.36
(S)-(-)-2, 2'-Diamino-1, 1'-binaphthalene
Off-white to beige
18531-95-8
100.0
Glass bottle
Powder
C20H16N2
MFCD00145204
≥98.5%

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.