Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-2,2'-Bis(N-diphenylphosphinoamino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, CTH-(S)-BINAM, 95%

Asymmetric hydrogenation

$102.99 - $394.05

Chemical Identifiers

Chemical Name or Material (S)- (-)-2,2'-Bis (N-diphenylphosphinoamino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthy
CAS 229177-79-1
Name Note CTH-(S)-BINAM
Molecular Formula C44H42N2P2
Formula Weight 660.78
MDL Number MFCD04974231
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4461902

  SDS
alfa aesar™
4461902
0.5g Each for $394.05

AA4461977

  SDS
alfa aesar™
4461977
0.1g Each for $102.99
Description & Specifications

Specifications

Formula Weight 660.78
Chemical Name or Material (S)- (-)-2,2'-Bis (N-diphenylphosphinoamino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthy
Name Note CTH-(S)-BINAM
Physical Form Powder
Assay 95%
Molecular Formula C44H42N2P2
MDL Number MFCD04974231
CAS 229177-79-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Packaged under Argon
Quantity 0.5g

  • Packaged under Argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Packaged under Argon