missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-2,2'-Bis(N-diphenylphosphinoamino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, CTH-(S)-BINAM, 95%

Asymmetric hydrogenation

Manufacturer:  Alfa Aesar™ 4461977

 View more versions of this product

Catalog No. AA4461977



Description

Description

  • Packaged under Argon
Specifications

Specifications

(S)- (-)-2,2'-Bis (N-diphenylphosphinoamino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthy
CTH-(S)-BINAM
C44H42N2P2
5,5'-bi 6-diphenylphosphinoamino tetralin
C1CCC2=C(C1)C=CC(=C2C3=C(C=CC4=C3CCCC4)NP(C5=CC=CC=C5)C6=CC=CC=C6)NP(C7=CC=CC=C7)C8=CC=CC=C8
660.782
660.78
95%
0.1g
229177-79-1
MFCD04974231
GTPWPBWUIWYZCB-UHFFFAOYSA-N
N-diphenylphosphanyl-1-[2-(diphenylphosphanylamino)-5,6,7,8-tetrahydronaphthalen-1-yl]-5,6,7,8-tetrahydronaphthalen-2-amine
10931066
Powder

Chemical Identifiers

229177-79-1
660.782
GTPWPBWUIWYZCB-UHFFFAOYSA-N
10931066
C1CCC2=C(C1)C=CC(=C2C3=C(C=CC4=C3CCCC4)NP(C5=CC=CC=C5)C6=CC=CC=C6)NP(C7=CC=CC=C7)C8=CC=CC=C8
C44H42N2P2
MFCD04974231
5,5'-bi 6-diphenylphosphinoamino tetralin
N-diphenylphosphanyl-1-[2-(diphenylphosphanylamino)-5,6,7,8-tetrahydronaphthalen-1-yl]-5,6,7,8-tetrahydronaphthalen-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

Documents